This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DMTF1 SIRNA Assays ›
Gene Symbol:
Dmtf1 LOC681224
Gene Name:
cyclin D binding myb-like transcription factor 1
similar to cyclin D binding myb-like transcription factor 1
Gene Aliases:
-
-
Chromosome Location:
Chr.4: 21862313 - 21902798 on Build Rnor_6.0
Chromosome Location:
Chr.18: 11599006 - 11601235 on Build Rnor_6.0
UniGene ID:
Rn.116028
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID 258872

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dmtf1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053693.1 NP_446145.1 17 2774
XM_006235993.1 XP_006236055.1 17 2836
GenBank mRNA BC081880.1 - 17 2774 7108159
Gene Symbol: LOC681224
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001060830.3 XP_001060830.1 1 1648

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
chromatin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0