This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C9ORF135 SIRNA Assays ›
Gene Symbol:
C9orf135
Gene Name:
chromosome 9 open reading frame 135
Gene Aliases:
-
Chromosome Location:
Chr.9: 69820804 - 69906237 on Build GRCh38
UniGene ID:
Hs.444459
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 258446

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C9orf135
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010940.2 NP_001010940.1 3 445
NM_001308084.1 NP_001295013.1 3 445
NM_001308085.1 NP_001295014.1 2 336
NM_001308086.1 NP_001295015.1 2 336
NR_131756.1 2 336
NR_131757.1 2 336
XM_011518229.2 XP_011516531.1 3 446
XM_011518230.2 XP_011516532.1 3 446
XM_011518231.2 XP_011516533.1 3 446
XM_011518233.2 XP_011516535.1 3 348
XM_011518234.2 XP_011516536.1 2 332
GenBank mRNA BC136709.1 - 3 422 9020699
BC144373.1 - 2 313
BC144374.1 - 2 313
BC144375.1 - 2 313
BC150564.1 - 3 423 40127624
BG182005.1 - 1 115
CD684431.1 - 3 473
CK126031.1 - 2 354

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0