This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CRYGB SIRNA Assays ›
Gene Symbol:
Crygb
Gene Name:
crystallin, gamma B
Gene Aliases:
Cryg-3, DGcry-3, Nop
Chromosome Location:
Chr.1: 65080222 - 65082290 on Build GRCm38
UniGene ID:
Mm.440053
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 257476

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Crygb
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144761.2 NP_658906.1 3 312
GenBank mRNA AK143110.1 - 3 311
BC056455.1 - 3 289 6824341
EF426303.1 - 3 297
K02585.1 - 2 167

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

eye development
lens development in camera-type eye
lens fiber cell development
lens fiber cell morphogenesis
structural constituent of eye lens

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0