This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZFP365 SIRNA Assays ›
Gene Symbol:
Zfp365
Gene Name:
zinc finger protein 365
Gene Aliases:
Znf365
Chromosome Location:
Chr.20: 22060160 - 22083766 on Build Rnor_6.0
UniGene ID:
Rn.24356
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 257390

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp365
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025145.1 NP_001020316.1 5 2584
GenBank mRNA BC087056.1 - 5 2584 7124000

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_178679, BC157991, BC080272, AK031846, BC157983, and others.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0