This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC10A3 SIRNA Assays ›
Gene Symbol:
Slc10a3
Gene Name:
solute carrier family 10, member 3
Gene Aliases:
-
Chromosome Location:
Chr.X: 156335711 - 156340256 on Build Rnor_6.0
UniGene ID:
Rn.140462
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 257281

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc10a3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024368.1 NP_001019539.1 3 558
XM_006229606.2 XP_006229668.1 3 1315
XM_008773611.1 XP_008771833.1 3 698
GenBank mRNA BC086556.1 - 3 558 7304843

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

response to organic substance
bile acid and bile salt transport
response to retinoic acid
transmembrane transport
bile acid:sodium symporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0