This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TUBA8 SIRNA Assays ›
Gene Symbol:
Tuba8
Gene Name:
tubulin, alpha 8
Gene Aliases:
-
Chromosome Location:
Chr.4: 153774511 - 153791223 on Build Rnor_6.0
UniGene ID:
Rn.27364
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 257196

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tuba8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024339.1 NP_001019510.1 2 156
XM_008763254.1 XP_008761476.1 2 293
GenBank mRNA BC079185.1 - 2 156 7131106
FQ217798.1 - 2 212

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

microtubule-based process
metabolic process
protein polymerization
GTPase activity
structural constituent of cytoskeleton
GTP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0