This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RAB20 SIRNA Assays ›
Gene Symbol:
RAB20
Gene Name:
RAB20, member RAS oncogene family
Gene Aliases:
-
Chromosome Location:
Chr.13: 110523066 - 110561737 on Build GRCh38
UniGene ID:
Hs.743563
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 25662

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RAB20
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017817.2 NP_060287.1 2 1011
GenBank mRNA AK000436.1 - 2 998
AK000446.1 - 2 998
BC026025.1 - 2 1007 4636491

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

small GTPase mediated signal transduction
protein transport
phagosome acidification
phagosome-lysosome fusion
regulation of autophagosome assembly
GTP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0