This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CAPN13 SIRNA Assays ›
Gene Symbol:
Capn13
Gene Name:
calpain 13
Gene Aliases:
CANP 13
Chromosome Location:
Chr.6: 24579590 - 24676533 on Build Rnor_6.0
UniGene ID:
Rn.15541
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 256480

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Capn13
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025133.1 NP_001020304.1 4 433
XM_006239777.2 XP_006239839.1 5 591
XM_006239778.2 XP_006239840.1 4 544
XM_006239779.2 XP_006239841.1 5 587
GenBank mRNA BC091250.1 - 4 433 7368359

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
calcium-dependent cysteine-type endopeptidase activity
calcium ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0