This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FOCAD SIRNA Assays ›
Gene Symbol:
FOCAD
Gene Name:
focadhesin
Gene Aliases:
KIAA1797
Chromosome Location:
Chr.9: 20658309 - 20995955 on Build GRCh38
UniGene ID:
Hs.136247
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 25645

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FOCAD
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017794.4 NP_060264.4 14, 15 1932
XM_005251494.3 XP_005251551.2 12, 13 1916
XM_011517945.2 XP_011516247.1 11, 12 1520
XM_017014852.1 XP_016870341.1 13, 14 1706
XM_017014854.1 XP_016870343.1 12, 13 1601
XM_017014855.1 XP_016870344.1 13, 14 1805
XM_017014856.1 XP_016870345.1 13, 14 1746
XM_017014858.1 XP_016870347.1 11, 12 1811
XM_017014859.1 XP_016870348.1 8, 9 1352
GenBank mRNA AK056522.1 - 14, 15 1932
AY139834.1 - 14, 15 1931
DB288580.1 - 2, 3 320

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0