This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LRRC56 SIRNA Assays ›
Gene Symbol:
Lrrc56
Gene Name:
leucine rich repeat containing 56
Gene Aliases:
RGD1311654
Chromosome Location:
Chr.1: 214181751 - 214197184 on Build Rnor_6.0
UniGene ID:
Rn.87507
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 256423

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lrrc56
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024902.1 NP_001020073.1 7, 8 793
XM_006230564.2 XP_006230626.1 7, 8 1279
XM_006230565.2 XP_006230627.1 7, 8 1389
XM_006230566.2 XP_006230628.1 7, 8 1562
XM_006230569.2 XP_006230631.1 7, 8 1316
XM_006230570.2 XP_006230632.1 5, 6 526
XM_008760005.1 XP_008758227.1 7, 8 1561
GenBank mRNA BC097444.1 - 7, 8 793 7456428

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0