This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BTBD16 SIRNA Assays ›
Gene Symbol:
Btbd16
Gene Name:
BTB (POZ) domain containing 16
Gene Aliases:
RGD1311454
Chromosome Location:
Chr.1: 201337363 - 201391246 on Build Rnor_6.0
UniGene ID:
Rn.137100
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 256275

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Btbd16
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017464.2 NP_001017464.1 5, 6 625
XM_006230403.2 XP_006230465.1 5, 6 725
XM_008759903.1 XP_008758125.1 5, 6 725
XM_008759904.1 XP_008758126.1 3, 4 391
XM_008759905.1 XP_008758127.1 5, 6 725
XM_008759906.1 XP_008758128.1 5, 6 725
GenBank mRNA BC079317.1 - 5, 6 628 7135047

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0