This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LRRFIP1 SIRNA Assays ›
Gene Symbol:
Lrrfip1
Gene Name:
leucine rich repeat (in FLII) interacting protein 1
Gene Aliases:
-
Chromosome Location:
Chr.9: 98139485 - 98268148 on Build Rnor_6.0
UniGene ID:
Rn.153995
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 256147

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lrrfip1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014269.1 NP_001014291.1 8 3143
XM_006245444.2 XP_006245506.1 22 3933
XM_006245445.2 XP_006245507.1 22 4090
XM_006245446.2 XP_006245508.1 21 3888
XM_006245447.2 XP_006245509.1 21 3861
XM_006245448.2 XP_006245510.1 21 3840
XM_006245450.2 XP_006245512.1 20 3746
XM_006245451.2 XP_006245513.1 19 3743
XM_006245452.2 XP_006245514.1 19 3674
XM_006245455.2 XP_006245517.1 11 3336
XM_006245456.2 XP_006245518.1 11 3494
XM_006245457.2 XP_006245519.1 10 3422
XM_006245458.2 XP_006245520.1 8 3078
XM_008767285.1 XP_008765507.1 21 3894
XM_008767286.1 XP_008765508.1 12 3414
GenBank mRNA BC081883.1 - 8 3143 7108398

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001111311, AK134120, AK162191, AK133661.

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0