This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SELT SIRNA Assays ›
Gene Symbol:
Selt
Gene Name:
selenoprotein T
Gene Aliases:
-
Chromosome Location:
Chr.2: 148765412 - 148781080 on Build Rnor_6.0
UniGene ID:
Rn.18892
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 256097

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Selt
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014253.2 NP_001014275.2 4 499
GenBank mRNA AY995234.1 - 4 499
BC086953.1 - 4 393 7321890
FQ211813.1 - 517

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_016275, BC026350, BC008411, AF131856, BC009611, and others.
This siRNA also targets the following transcripts in Mouse: NM_001040396, AK146778, AK032838, BC132453, BC038867, and others.

Gene Ontology Categories:

Function(s) Process(es)

response to glucose
pancreas development
insulin secretion involved in cellular response to glucose stimulus
glucose homeostasis
cell redox homeostasis
selenium binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0