This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other STK19 SIRNA Assays ›
Gene Symbol:
Stk19 LOC103694374
Gene Name:
serine/threonine kinase 19
serine/threonine-protein kinase 19
Gene Aliases:
-
-
Chromosome Location:
Chr.20: 4317440 - 4517128 on Build Rnor_6.0
Chromosome Location:
Chr.20: 2665615 - 2666517 on Build Rnor_6.0
UniGene ID:
Rn.17283
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID 255314

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Stk19
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013197.2 NP_001013215.2 7 1102
GenBank mRNA BC061571.1 - 7 1106 6921289
Gene Symbol: LOC103694374
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008772703.1 XP_008770925.1 1105

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0