This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other COA6 SIRNA Assays ›
Gene Symbol:
COA6
Gene Name:
cytochrome c oxidase assembly factor 6
Gene Aliases:
C1orf31, CEMCOX4
Chromosome Location:
Chr.1: 234373437 - 234384049 on Build GRCh38
UniGene ID:
Hs.23198
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 255240

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: COA6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012985.2 NP_001013003.1 3 529
NM_001206641.2 NP_001193570.2 3 613
NM_001301733.1 NP_001288662.1 2 900
GenBank mRNA BC025793.1 - 3 436
BC035925.1 - 3 526
BC116455.1 - 3 529 4540930
BG195363.1 - 1 216
BG204581.1 - 2 372
CB136291.1 - 3 582

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

respiratory chain complex IV assembly
plasma membrane ATP synthesis coupled electron transport
hydrogen ion transmembrane transport
cytochrome-c oxidase activity
copper ion binding
protein binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0