This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OLFR1137 SIRNA Assays ›
Gene Symbol:
Olfr1137
Gene Name:
olfactory receptor 1137
Gene Aliases:
MOR177-20, MOR40-9P
Chromosome Location:
Chr.2: 87710972 - 87711904 on Build GRCm38
UniGene ID:
Mm.377350
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 254901

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olfr1137
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011833.1 NP_001011833.1 1 250

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity
odorant binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0