This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OR52I2 SIRNA Assays ›
Gene Symbol:
OR52I2
Gene Name:
olfactory receptor family 52 subfamily I member 2
Gene Aliases:
OR11-12, OR52I1
Chromosome Location:
Chr.11: 4586791 - 4587905 on Build GRCh38
UniGene ID:
Hs.553607
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 254361

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: OR52I2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005170.2 NP_001005170.1 1 788
GenBank mRNA BC136921.1 - 1 788 9020912
BC151144.1 - 1 788 9020913

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0