This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AI413582 SIRNA Assays ›
Gene Symbol:
AI413582
Gene Name:
expressed sequence AI413582
Gene Aliases:
AI854251
Chromosome Location:
Chr.17: 27563766 - 27565799 on Build GRCm38
UniGene ID:
Mm.61488
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 254192

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: AI413582
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002895.2 NP_001002895.2 4 324
GenBank mRNA AK131593.1 - 4 319
AK131681.1 - 4 299
BC028464.1 - 4 297 1264774
BY244090.1 - 3 476

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0