This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other METTL15 SIRNA Assays ›
Gene Symbol:
Mettl15
Gene Name:
methyltransferase like 15
Gene Aliases:
RGD1304978
Chromosome Location:
Chr.3: 100186106 - 100366148 on Build Rnor_6.0
UniGene ID:
Rn.160141
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 253111

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mettl15
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024981.1 NP_001020152.1 2, 3 315
XM_006234686.2 XP_006234748.1 3, 4 487
XM_006234687.2 XP_006234749.1 2, 3 386
XM_006234688.2 XP_006234750.1 3, 4 482
XM_006234689.2 XP_006234751.1 3, 4 461
XM_008762084.1 XP_008760306.1 3, 4 570
XM_008762085.1 XP_008760307.1 3, 4 567
XM_008762086.1 XP_008760308.1 3, 4 549
XM_008762087.1 XP_008760309.1 3, 4 566
GenBank mRNA BC091215.1 - 2, 3 315 7383613

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_029790, BC106186, BC115926, AK002682, BC117549.

Gene Ontology Categories:

Function(s) Process(es)

rRNA base methylation
rRNA (cytosine-N4-)-methyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0