This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM5 SIRNA Assays ›
Gene Symbol:
Tmem5
Gene Name:
transmembrane protein 5
Gene Aliases:
-
Chromosome Location:
Chr.7: 64329341 - 64341196 on Build Rnor_6.0
UniGene ID:
Rn.161056
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 253013

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024759.1 NP_001019930.1 6 1359
XM_006241397.2 XP_006241459.1 6 1424
XM_008765391.1 XP_008763613.1 5 1111
GenBank mRNA BC097355.1 - 6 1359 7441970

Back To Top

More Information


Gene Ontology Categories:

Function(s)

protein O-linked mannosylation

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm