This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DDX47 SIRNA Assays ›
Gene Symbol:
Ddx47
Gene Name:
DEAD (Asp-Glu-Ala-Asp) box polypeptide 47
Gene Aliases:
-
Chromosome Location:
Chr.4: 168772558 - 168787594 on Build Rnor_6.0
UniGene ID:
Rn.73790
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 252851

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ddx47
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001015005.2 NP_001015005.2 8 835
XM_006237518.1 XP_006237580.1 9 892
GenBank mRNA BC091696.1 - 8 828 7301732

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026360, AK169595, BC095944, AK076982, BC006843, and others.

Gene Ontology Categories:

Function(s) Process(es)

rRNA processing
RNA splicing
extrinsic apoptotic signaling pathway via death domain receptors
helicase activity
ATP binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0