This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ANKRD46 SIRNA Assays ›
Gene Symbol:
Ankrd46
Gene Name:
ankyrin repeat domain 46
Gene Aliases:
RGD1309382, ank-s
Chromosome Location:
Chr.7: 75279443 - 75300430 on Build Rnor_6.0
UniGene ID:
Rn.11978
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 252738

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ankrd46
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013948.1 NP_001013970.1 5 1453
XM_006241530.2 XP_006241592.1 5 1724
XM_006241531.2 XP_006241593.1 4 1356
XM_006241532.2 XP_006241594.1 4 3097
XM_006241533.2 XP_006241595.1 4 3023
GenBank mRNA AB095364.1 - 5 1453
BC105616.1 - 5 1449 7453107

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0