This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRAPPC13 SIRNA Assays ›
Gene Symbol:
Trappc13
Gene Name:
trafficking protein particle complex 13
Gene Aliases:
RGD1306583
Chromosome Location:
Chr.2: 34223396 - 34254988 on Build Rnor_6.0
UniGene ID:
Rn.15285
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 252667

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Trappc13
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013908.2 NP_001013930.2 5 682
XM_006231884.2 XP_006231946.1 5 721
XM_006231885.2 XP_006231947.1 5 721
XM_006231886.2 XP_006231948.1 5 721
GenBank mRNA BC088172.1 - 5 656 7323103
DY312499.1 - 5 682
FQ217190.1 - 665

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0