siRNA Details
Gene Symbol: LOC288521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013868.1 | NP_001013890.1 | 1164 | |||
GenBank mRNA | BC079301.1 | - | 1164 | 7134538 |

Gene Symbol: LOC288521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013868.1 | NP_001013890.1 | 1164 | |||
GenBank mRNA | BC079301.1 | - | 1164 | 7134538 |