This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other THEG SIRNA Assays ›
Gene Symbol:
Theg
Gene Name:
theg spermatid protein
Gene Aliases:
-
Chromosome Location:
Chr.7: 13013521 - 13023666 on Build Rnor_6.0
UniGene ID:
Rn.49140
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 252473

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Theg
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013102.1 NP_001013120.1 7 884
XM_006240857.2 XP_006240919.1 7 904
XM_006240858.2 XP_006240920.1 6 832
GenBank mRNA BC083921.1 - 7 884 7135133

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_199226, NM_011583, AJ011834, AB033128, AK017058, and others.

Gene Ontology Categories:

Function(s)

multicellular organism development
spermatogenesis
cell differentiation

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0