This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TERF1 SIRNA Assays ›
Gene Symbol:
Terf1
Gene Name:
telomeric repeat binding factor (NIMA-interacting) 1
Gene Aliases:
-
Chromosome Location:
Chr.5: 2853455 - 2882728 on Build Rnor_6.0
UniGene ID:
Rn.33853
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 252304

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Terf1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012464.1 NP_001012482.1 2 318
XM_006237729.2 XP_006237791.1 2 378
GenBank mRNA BC089888.1 - 2 318 7374057

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G2/M transition of mitotic cell cycle
obsolete age-dependent telomere shortening
telomere maintenance via telomerase
mitotic spindle assembly checkpoint
negative regulation of DNA replication
obsolete telomere maintenance via telomere shortening
positive regulation of microtubule polymerization
telomeric loop formation
negative regulation of telomere maintenance via telomerase
response to drug
positive regulation of apoptotic process
positive regulation of mitotic nuclear division
positive regulation of mitotic cell cycle
protein homooligomerization
negative regulation of telomerase activity
molecular_function
chromatin binding
double-stranded telomeric DNA binding
microtubule binding
DNA binding, bending
telomeric DNA binding
protein homodimerization activity
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0