This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NDUFS2 SIRNA Assays ›
Gene Symbol:
Ndufs2
Gene Name:
NADH dehydrogenase (ubiquinone) Fe-S protein 2
Gene Aliases:
-
Chromosome Location:
Chr.13: 89606848 - 89623419 on Build Rnor_6.0
UniGene ID:
Rn.99666
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 252010

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ndufs2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011907.1 NP_001011907.1 9 1110
XM_003751277.2 XP_003751325.1 9 1123
GenBank mRNA BC082067.1 - 9 1110 7132777
FQ215922.1 - 9 1114

Back To Top

More Information


Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

response to oxidative stress
biological_process
oxidation-reduction process
molecular_function
NADH dehydrogenase activity
NADH dehydrogenase (ubiquinone) activity
metal ion binding
quinone binding
NAD binding
4 iron, 4 sulfur cluster binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0