This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC10A7 SIRNA Assays ›
Gene Symbol:
Slc10a7
Gene Name:
solute carrier family 10, member 7
Gene Aliases:
P7
Chromosome Location:
Chr.19: 32857984 - 33080772 on Build Rnor_6.0
UniGene ID:
Rn.23101
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 251936

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc10a7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010948.1 NP_001010948.1 11 893
GenBank mRNA AY825929.1 - 11 893
FQ211514.1 - 1081

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001009981, NM_029736, NM_001282109, NM_001282108, AY825928, and others.

Gene Ontology Categories:

Function(s) Process(es)

sodium ion transport
transmembrane transport
symporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0