This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF581 SIRNA Assays ›
Gene Symbol:
ZNF581
Gene Name:
zinc finger protein 581
Gene Aliases:
HSPC189
Chromosome Location:
Chr.19: 55635028 - 55645623 on Build GRCh38
UniGene ID:
Hs.20768
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 24689

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF581
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016535.3 NP_057619.1 2 463
XM_006723240.3 XP_006723303.1 2 486
XM_017026867.1 XP_016882356.1 2 396
GenBank mRNA AF151023.2 - 2 463
AK026203.1 - 3 400
AK315603.1 - 2 434
BC001219.1 - 2 432 3528677
BC010097.2 - 2 435 3504825
BC071620.1 - 2 562 4446945
BC071845.1 - 2 417 4764567

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0