This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PRRT3 SIRNA Assays ›
Gene Symbol:
PRRT3
Gene Name:
proline rich transmembrane protein 3
Gene Aliases:
-
Chromosome Location:
Chr.3: 9945542 - 9952415 on Build GRCh38
UniGene ID:
Hs.598147
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 242255

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PRRT3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318871.1 NP_001305800.1 2 402
NM_207351.4 NP_997234.3 2 402
NR_134900.1 2 402
XM_011533628.2 XP_011531930.1 2 480
XM_011533629.2 XP_011531931.1 2 460
XM_017006256.1 XP_016861745.1 1 2267
GenBank mRNA AK090993.1 - 2 381
AK296317.1 - 2 364
AY358158.1 - 2 389
BC040508.1 - 2 381 5300209
BC089447.1 - 2 361 30563721
BC111555.1 - 1 299
BM544786.1 - 2 426
DA198780.1 - 2 381

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0