This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC39A14 SIRNA Assays ›
Gene Symbol:
SLC39A14
Gene Name:
solute carrier family 39 member 14
Gene Aliases:
HMNDYT2, LZT-Hs4, NET34, ZIP14, cig19
Chromosome Location:
Chr.8: 22367249 - 22434129 on Build GRCh38
UniGene ID:
Hs.491232
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 241612

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SLC39A14
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128431.2 NP_001121903.1 2 318
NM_001135153.1 NP_001128625.1 2 359
NM_001135154.1 NP_001128626.1 2 318
NM_015359.4 NP_056174.2 2 318
XM_005273465.2 XP_005273522.1 3 406
XM_005273466.4 XP_005273523.1 2 390
XM_006716323.2 XP_006716386.1 4 604
XM_006716324.2 XP_006716387.1 3 395
XM_011544478.2 XP_011542780.1 3 470
XM_017013292.1 XP_016868781.1 3 486
XM_017013293.1 XP_016868782.1 4 604
XM_017013294.1 XP_016868783.1 4 513
XM_017013295.1 XP_016868784.1 2 523
GenBank mRNA AK172810.1 - 2 289
AK295807.1 - 2 234
BC015770.1 - 2 280 4865469
D31887.1 - 2 262
DC323208.1 - 2 318
DC423882.1 - 2 360
KU178568.1 - 1 143
KU178569.1 - 1 143
KU178570.1 - 1 143
KU178571.1 - 1 143

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cellular zinc ion homeostasis
zinc II ion transmembrane transport
zinc II ion transmembrane import
ferrous iron transmembrane transport
zinc ion transmembrane transporter activity
ferrous iron transmembrane transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0