This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KCTD3 SIRNA Assays ›
Gene Symbol:
KCTD3
Gene Name:
potassium channel tetramerization domain containing 3
Gene Aliases:
NY-REN-45
Chromosome Location:
Chr.1: 215567185 - 215621821 on Build GRCh38
UniGene ID:
Hs.335139
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 23810

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KCTD3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319294.1 NP_001306223.1 5 596
NM_001319295.1 NP_001306224.1 5 596
NM_016121.4 NP_057205.2 5 596
XM_005273158.1 XP_005273215.1 5 790
GenBank mRNA AK296275.1 - 3 129
BC013868.2 - 5 583
BC094879.1 - 5 545 3078971
BC117188.1 - 5 478 40125739
BC126173.1 - 5 478 8991889

Back To Top

More Information


Gene Ontology Categories:

Function(s)

protein homooligomerization

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0