This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZDHHC9 SIRNA Assays ›
Gene Symbol:
ZDHHC9
Gene Name:
zinc finger DHHC-type containing 9
Gene Aliases:
CGI89, CXorf11, DHHC9, MMSA1, MRXSZ, ZDHHC10, ZNF379, ZNF380
Chromosome Location:
Chr.X: 129803288 - 129843934 on Build GRCh38
UniGene ID:
Hs.193566
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 23742

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZDHHC9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008222.2 NP_001008223.1 10 1621
NM_016032.3 NP_057116.2 11 1601
GenBank mRNA AB209807.1 - 10 1540
AF151847.1 - 11 1489
AK001112.1 - 11 1551
AK001424.1 - 8 935
AK001524.1 - 10 1352
AK096967.1 - 11 1252
AL161962.1 - 11 1478
AY358558.1 - 11 1533
AY952881.8 - 11 1573
BC000035.2 - 10 1495 3506601
BC003128.1 - 11 1539 2957911
BC006200.2 - 11 1535 2964425
BC012826.2 - 11 1535 2964425

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

peptidyl-L-cysteine S-palmitoylation
protein palmitoylation
zinc ion binding
palmitoyltransferase activity
protein-cysteine S-palmitoyltransferase activity
Ras palmitoyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0