This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EDRF1 SIRNA Assays ›
Gene Symbol:
EDRF1
Gene Name:
erythroid differentiation regulatory factor 1
Gene Aliases:
C10orf137
Chromosome Location:
Chr.10: 125719515 - 125764143 on Build GRCh38
UniGene ID:
Hs.468688
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 23386

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: EDRF1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001202438.1 NP_001189367.1 19 2956
NM_015608.2 NP_056423.2 18 2854
NR_110857.1 19 2951
NR_110858.1 20 3082
NR_110859.1 19 2972
XM_011539622.1 XP_011537924.1 16 2144
XM_017016066.1 XP_016871555.1 19 2700
GenBank mRNA AK091507.1 - 18 2666
AK314958.1 - 4 690
AL050102.1 - 3 432
BC016903.1 - 7 952
BC026172.1 - 20 3083 4838438
BC105929.1 - 19 2890
BC107479.1 - 19 2895 40028896

Back To Top

More Information


Gene Ontology Categories:

Function(s)

transcription, DNA-templated
regulation of transcription, DNA-templated

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0