This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FNBP4 SIRNA Assays ›
Gene Symbol:
FNBP4
Gene Name:
formin binding protein 4
Gene Aliases:
FBP30
Chromosome Location:
Chr.11: 47716510 - 47767478 on Build GRCh38
UniGene ID:
Hs.6834
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 22767

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FNBP4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318339.1 NP_001305268.1 2 447
NM_015308.3 NP_056123.2 2 441
GenBank mRNA AB023231.2 - 2 440
AK054636.1 - 2 264
AK123898.1 - 2 245
AL137480.1 - 2 240
BC037404.1 - 2 272 5140963
DA238225.1 - 2 429
DB458472.1 - 2 423

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0