This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FNBP4 SIRNA Assays ›
Gene Symbol:
FNBP4
Gene Name:
formin binding protein 4
Gene Aliases:
FBP30
Chromosome Location:
Chr.11: 47716510 - 47767478 on Build GRCh38
UniGene ID:
Hs.6834
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 22671

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FNBP4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318339.1 NP_001305268.1 2 444
NM_015308.3 NP_056123.2 2 438
GenBank mRNA AB023231.2 - 2 437
AK054636.1 - 2 261
AK123898.1 - 2 242
AL137480.1 - 2 237
BC037404.1 - 2 269 5140963
DA238225.1 - 2 426
DB458472.1 - 2 420

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0