This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CHDH SIRNA Assays ›
Gene Symbol:
Chdh
Gene Name:
choline dehydrogenase
Gene Aliases:
CDH, CHD
Chromosome Location:
Chr.16: 6078086 - 6109386 on Build Rnor_6.0
UniGene ID:
Rn.22857
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 224052

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Chdh
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198731.2 NP_942026.1 8 2987
XM_006252578.2 XP_006252640.1 8 3052
XM_006252579.2 XP_006252641.1 8 3006
XM_006252580.2 XP_006252642.1 8 2812
XM_008770984.1 XP_008769206.1 8 3355
XM_008770985.1 XP_008769207.1 9 3444
GenBank mRNA BC085787.1 - 8 2987 7108667

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

glycine betaine biosynthetic process from choline
oxidation-reduction process
choline dehydrogenase activity
flavin adenine dinucleotide binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0