This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZPBP2 SIRNA Assays ›
Gene Symbol:
Zpbp2
Gene Name:
zona pellucida binding protein 2
Gene Aliases:
1700017D11Rik, 2610022C02Rik, ZPBPL
Chromosome Location:
Chr.11: 98551097 - 98558665 on Build GRCm38
UniGene ID:
Mm.55996
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 223899

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zpbp2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166494.1 NP_001159966.1 6 1086
NM_001166495.1 NP_001159967.1 7 875
NM_027061.2 NP_081337.2 7 966
NM_199419.3 NP_955451.2 6 747
GenBank mRNA AK006058.1 - 6 1034
AY251601.1 - 7 911
AY251602.1 - 6 692
BC050986.1 - 7 842
BC089476.1 - 6 747 6708062

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

acrosome assembly
binding of sperm to zona pellucida
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header