This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EHMT2 SIRNA Assays ›
Gene Symbol:
Ehmt2
Gene Name:
euchromatic histone lysine N-methyltransferase 2
Gene Aliases:
Bat8, G9a, Ng36
Chromosome Location:
Chr.20: 4576062 - 4592980 on Build Rnor_6.0
UniGene ID:
Rn.116518
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 223616

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ehmt2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_212463.1 NP_997628.1 23 3179
XM_006255954.2 XP_006256016.2 23 3214
XM_006255955.2 XP_006256017.2 22 3112
XM_006255956.2 XP_006256018.2 22 3081
XM_006255957.1 XP_006256019.1 24 3038
XM_006255958.1 XP_006256020.1 24 3008
XM_006255959.1 XP_006256021.1 23 2906

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001286575, NM_147151, NM_001286573, NM_145830, BC058357, and others.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
regulation of DNA replication
DNA methylation
regulation of transcription from RNA polymerase II promoter
synaptonemal complex assembly
germ cell development
spermatid development
long-term memory
fertilization
DNA methylation on cytosine within a CG sequence
histone methylation
peptidyl-lysine methylation
peptidyl-lysine dimethylation
response to nutrient levels
histone lysine methylation
organ growth
cellular response to drug
regulation of DNA methylation
response to ethanol
histone H3-K9 methylation
regulation of histone H3-K4 methylation
regulation of histone H3-K9 methylation
response to fungicide
histone H3-K27 methylation
RNA polymerase II regulatory region sequence-specific DNA binding
p53 binding
protein binding
zinc ion binding
protein-lysine N-methyltransferase activity
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H3-K9 specific)
histone methyltransferase activity (H3-K27 specific)
C2H2 zinc finger domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header