This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FBXO41 SIRNA Assays ›
Gene Symbol:
Fbxo41
Gene Name:
F-box protein 41
Gene Aliases:
9630017H13, D6Ertd538e
Chromosome Location:
Chr.6: 85469574 - 85503028 on Build GRCm38
UniGene ID:
Mm.38777
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 223079

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fbxo41
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001160.3 NP_001001160.1 14 3738
NM_001289674.1 NP_001276603.1 13 3569
NM_001289675.1 NP_001276604.1 13 3563
GenBank mRNA AK035911.1 - 2 673
AK129466.1 - 12 3115
AK147230.1 - 14 3630
AK147681.1 - 13 3428
BC061475.1 - 11 2210
BC070445.1 - 13 3569 30536738

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0