This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OLR1252 SIRNA Assays ›
Gene Symbol:
Olr1252 LOC100909736 LOC100912507 LOC103692018
Gene Name:
olfactory receptor 1252
olfactory receptor 8B3-like
olfactory receptor 8B3-like
olfactory receptor 8B3-like
Gene Aliases:
-
-
-
-
Chromosome Location:
Chr.8: 42364887 - 42365819 on Build Rnor_6.0
Chromosome Location:
Chr.8: 42360580 - 42361272 on Build Rnor_6.0
Chromosome Location:
Chr.3: 158919862 - 158920794 on Build Rnor_6.0
Chromosome Location:
Chr.4: 1685913 - 1686845 on Build Rnor_6.0
UniGene ID:
Rn.143518
-
-
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
-
-
siRNA ID 222036

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olr1252
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000454.1 NP_001000454.1 1 437
Gene Symbol: LOC100909736
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750485.1 XP_003750533.1 248
Gene Symbol: LOC100912507
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008762440.1 XP_008760662.1 1 437
Gene Symbol: LOC103692018
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008762660.1 XP_008760882.1 1 437

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
detection of chemical stimulus involved in sensory perception of smell
olfactory receptor activity
G-protein coupled receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0