This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OLR56 SIRNA Assays ›
Gene Symbol:
Olr56 LOC103690133
Gene Name:
olfactory receptor 56
olfactory receptor 51F2-like
Gene Aliases:
-
-
Chromosome Location:
Chr.1: 167944448 - 167945398 on Build Rnor_6.0
Chromosome Location:
Chr.1: 168061722 - 168069717 on Build Rnor_6.0
UniGene ID:
Rn.141921
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID 221658

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olr56
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000130.1 NP_001000130.1 1 398
Gene Symbol: LOC103690133
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759842.1 XP_008758064.1 1 80

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
detection of chemical stimulus involved in sensory perception of smell
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-77574795ff-5hfml:80/100.66.128.142:80.
git-commit: 5cc8a939e1477954742eeb281a87e54f28adfb77
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header