This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNF123 SIRNA Assays ›
Gene Symbol:
Rnf123
Gene Name:
ring finger protein 123
Gene Aliases:
BC003945, Kpc1
Chromosome Location:
Chr.9: 108051672 - 108079773 on Build GRCm38
UniGene ID:
Mm.379272
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 221469

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rnf123
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032543.2 NP_115932.1 4 381
GenBank mRNA AK137599.1 - 4 326
AK157870.1 - 4 291
AK158320.1 - 4 333
AK172244.1 - 4 305
AY744153.1 - 3 180
BC025530.1 - 4 302
BC057082.1 - 4 776 5704926
BY286344.1 - 4 308

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0