This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BCAP31 SIRNA Assays ›
Gene Symbol:
Bcap31
Gene Name:
B-cell receptor-associated protein 31
Gene Aliases:
Bap31
Chromosome Location:
Chr.X: 157094444 - 157126397 on Build Rnor_6.0
UniGene ID:
Rn.96419
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 220521

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Bcap31
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004224.1 NP_001004224.1 5 552
XM_006229556.2 XP_006229618.1 4 509
XM_006229557.2 XP_006229619.1 5 626
GenBank mRNA BC079182.1 - 5 552 7130880
FQ219566.1 - 5 543
FQ219790.1 - 5 551
FQ230132.1 - 5 543
FQ230136.1 - 5 543

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_012060, BC002106, AK011500, AK008043, AK151641, and others.

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
ER to Golgi vesicle-mediated transport
immune response
positive regulation of cytosolic calcium ion concentration
spermatogenesis
biological_process
negative regulation of endoplasmic reticulum calcium ion concentration
calcium-mediated signaling using intracellular calcium source
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of mitochondrial calcium ion concentration
protein localization to endoplasmic reticulum exit site
positive regulation of intrinsic apoptotic signaling pathway
protein binding
protein complex binding
MHC class I protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0