This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KATNA1 SIRNA Assays ›
Gene Symbol:
Katna1
Gene Name:
katanin p60 (ATPase-containing) subunit A1
Gene Aliases:
-
Chromosome Location:
Chr.1: 1826170 - 1867786 on Build Rnor_6.0
UniGene ID:
Rn.47464
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 220503

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Katna1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004217.4 NP_001004217.2 11 1577
GenBank mRNA AY621629.1 - 10 1381
BC097929.1 - 11 1577 7444887

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011835, AV135581, AF153197, AK012319, AK041900, and others.

Gene Ontology Categories:

Function(s) Process(es)

microtubule bundle formation
neuron migration
mitotic nuclear division
protein localization
metabolic process
negative regulation of neuron projection development
protein ubiquitination
cytoplasmic microtubule organization
microtubule severing
cell division
ubiquitin-protein transferase activity
ATP binding
microtubule binding
microtubule-severing ATPase activity
ATPase activity
dynein binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0