This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC5A12 SIRNA Assays ›
Gene Symbol:
Slc5a12
Gene Name:
solute carrier family 5 (sodium/glucose cotransporter), member 12
Gene Aliases:
AI315119, D630015G19, SMCT2
Chromosome Location:
Chr.2: 110597299 - 110649346 on Build GRCm38
UniGene ID:
Mm.277148
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 220136

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc5a12
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003915.2 NP_001003915.1 15 3318
NM_001177624.1 NP_001171095.1 15 3306
GenBank mRNA AF529222.1 - 15 3326
AK085356.1 - 15 3301
AK085582.1 - 2 1357
AK128951.1 - 15 3298
AY964639.1 - 15 3009

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
sodium ion transport
transmembrane transport
transporter activity
symporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0