This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNF144A SIRNA Assays ›
Gene Symbol:
RNF144A
Gene Name:
ring finger protein 144A
Gene Aliases:
RNF144, UBCE7IP4
Chromosome Location:
Chr.2: 6917392 - 7076886 on Build GRCh38
UniGene ID:
Hs.22146
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 22007

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RNF144A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014746.4 NP_055561.2 9 2694
XM_005246200.3 XP_005246257.1 9 3885
XM_005246202.4 XP_005246259.1 8 2525
XM_017005396.1 XP_016860885.1 9 3861
XM_017005397.1 XP_016860886.1 9 2519
XM_017005404.1 XP_016860893.1 8 2502
GenBank mRNA BQ423668.1 - 1 592
D79983.1 - 9 2600

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
zinc ion binding
ligase activity
ubiquitin conjugating enzyme binding
ubiquitin protein ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header