This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CCNT2 SIRNA Assays ›
Gene Symbol:
Ccnt2
Gene Name:
cyclin T2
Gene Aliases:
2900041I18Rik, C81304, CycT2, CycT2a, CycT2b
Chromosome Location:
Chr.1: 127774164 - 127804837 on Build GRCm38
UniGene ID:
Mm.268672
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 219638

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ccnt2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028399.1 NP_082675.1 9 2171
GenBank mRNA AK036772.1 - 2021
AK047125.1 - 5 3071
AK049863.1 - 10 2018
AK152399.1 - 9 2119
AK154439.1 - 9 2129
AK156231.1 - 2042
BC054122.1 - 9 2171 6509024
JX965958.1 - 9 2107

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001241, NM_058241, AF048732, AK292913, BC114366, and others.
This siRNA also targets the following transcripts in Rat: NM_001107171, XM_008769457, XM_006249686, XM_006249687, XM_008769458.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0