This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM160B1 SIRNA Assays ›
Gene Symbol:
FAM160B1
Gene Name:
family with sequence similarity 160 member B1
Gene Aliases:
KIAA1600, bA106M7.3
Chromosome Location:
Chr.10: 114821744 - 114899827 on Build GRCh38
UniGene ID:
Hs.192619
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 219590

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM160B1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135051.1 NP_001128523.1 14, 15 2290
NM_020940.3 NP_065991.3 14, 15 2290
GenBank mRNA AB046820.2 - 13, 14 1885
AL833226.1 - 3, 4 349
BC037207.1 - 14, 15 2290 4828347
CR933692.1 - 14, 15 2010

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0