This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CCNB2 SIRNA Assays ›
Gene Symbol:
Ccnb2 LOC100364016
Gene Name:
cyclin B2
cyclin B2-like
Gene Aliases:
-
-
Chromosome Location:
Chr.8: 76847972 - 76861165 on Build Rnor_6.0
Chromosome Location:
Chr.1: 21145363 - 21146880 on Build Rnor_6.0
UniGene ID:
Rn.6743
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID 219250

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ccnb2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009470.1 NP_001009470.1 3 259
GenBank mRNA BC088212.1 - 3 259 7383459
BC097952.1 - 3 283 7454811
FQ228772.1 - 3 273
FQ234119.1 - 3 273
Gene Symbol: LOC100364016
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008758651.1 XP_008756873.1 1 293

Back To Top

More Information


Gene Ontology Categories:

Function(s)

in utero embryonic development
growth
T cell homeostasis
thymus development
cell division

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0